Anti-IGLL1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
Proveedor: Bioss
BS-15577R-A647
BOSSBS-15577R-A647EA 893 EUR
BOSSBS-15577R-A647
Anti-IGLL1 Rabbit Polyclonal Antibody (ALEXA FLUOR® 647)
Anticuerpos
The preB cell receptor is found on the surface of proB and preB cells, where it is involved in transduction of signals for cellular proliferation, differentiation from the proB cell to the preB cell stage, allelic exclusion at the Ig heavy chain gene locus, and promotion of Ig light chain gene rearrangements. The preB cell receptor is composed of a membrane-bound Ig mu heavy chain in association with a heterodimeric surrogate light chain. This gene encodes one of the surrogate light chain subunits and is a member of the immunoglobulin gene superfamily. This gene does not undergoMore Product Information