Anti-APOH Rabbit Polyclonal Antibody (Cy7®)

Proveedor: Bioss
BS-1570R-CY7
BOSSBS-1570R-CY7EA 900 EUR
BOSSBS-1570R-CY7
Anti-APOH Rabbit Polyclonal Antibody (Cy7®)
Anticuerpos
Binds to various kinds of negatively charged substances such as heparin, phospholipids, and dextran sulfate. May prevent activation of the intrinsic blood coagulation cascade by binding to phospholipids on the surface of damaged cells.

Type: Primary
Antigen: APOH
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 281-345/345
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR