Anti-GAL3ST4 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)

Proveedor: Bioss
BS-16215R-A680
BOSSBS-16215R-A680EA 890 EUR
BOSSBS-16215R-A680
Anti-GAL3ST4 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Anticuerpos
Catalyses the transfer of sulfate to beta-1,3-linked galactose residues in O-linked glycoproteins. Good substrates include asialofetuin, Gal-beta-1,3-GalNAc and Gal-beta-1,3 (GlcNAc-beta-1,6)GalNAc.

Type: Primary
Antigen: GAL3ST4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-486/486
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR