Anti-fowl typhoid and pullorum disease Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Proveedor: Bioss
BS-1675R-A750
BOSSBS-1675R-A750EA 890 EUR
BOSSBS-1675R-A750
Anti-fowl typhoid and pullorum disease Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
Fowl typhoid and pullorum disease.

Type: Primary
Antigen: fowl typhoid and pullorum disease
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

ESPECIFICACIONES

Environmentally Preferable
Nombre del antígeno fowl typhoid and pullorum disease
Sinónimos de antígenos NULL
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 750
Reactividad
Huésped Rabbit
Isotipo IgG
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Modificación Unmodified
Inmunógeno 0
Forma Liquid
Purification Purified by Protein A
Concentración 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Temperatura de almacenamiento Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR