Anti-Peptide YY/PYY Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Bioss
BS-2265R-HRP
BOSSBS-2265R-HRPEA 900 EUR
BOSSBS-2265R-HRP
Anti-Peptide YY/PYY Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
This gut peptide inhibits exocrine pancreatic secretion, has a vasoconstrictory action and inhibitis jejunal and colonic mobility.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: Peptide YY/PYY
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR