Anti-EMC1 Rabbit Polyclonal Antibody (Cy5.5®)

Proveedor: Bioss
BS-2279R-CY5.5
BOSSBS-2279R-CY5.5EA 900 EUR
BOSSBS-2279R-CY5.5
Anti-EMC1 Rabbit Polyclonal Antibody (Cy5.5®)
Anticuerpos
This gene encodes a single-pass type I transmembrane protein, which is a subunit of the endoplasmic reticulum membrane protein complex (EMC). Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2012].

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: EMC1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR