Anti-CYP2B6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-2352R-A488
BOSSBS-2352R-A488EA 876 EUR
BOSSBS-2352R-A488
Anti-CYP2B6 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
Cytochromes P45 are a group of heme-thiolate monooxygenases. In liver microsomes, this enzyme is involved in an NADPH-dependent electron transport pathway. It oxidizes a variety of structurally unrelated compounds, including steroids, fatty acids, and xenobiotics. Acts as a 1,4-cineole 2-exo-monooxygenase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP2B6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR