Anti-FOXA2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Proveedor: Bioss
BS-2358R-A555
BOSSBS-2358R-A555EA 901 EUR
BOSSBS-2358R-A555
Anti-FOXA2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticuerpos
Transcription factor that is involved in embryonic development, establishment of tissue-specific gene expression and regulation of gene expression in differentiated tissues. Is thought to act as a 'pioneer' factor opening the compacted chromatin for other proteins through interactions with nucleosomal core histones and thereby replacing linker histones at target enhancer and/or promoter sites. Binds DNA with the consensus sequence 5'-[AC]A[AT]T[AG]TT[GT][AG][CT]T[CT]-3' (By similarity). In embryonic development is required for notochord formation. Involved in the development of multipleMore Product Information