Anti-CYP7A1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-2399R-A488
BOSSBS-2399R-A488EA 901 EUR
BOSSBS-2399R-A488
Anti-CYP7A1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
Catalyzes a rate-limiting step in cholesterol catabolism and bile acid biosynthesis by introducing a hydrophilic moiety at position 7 of cholesterol. Important for cholesterol homeostasis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP7A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR