Anti-TNFRSF9 Rabbit Polyclonal Antibody

Proveedor: Bioss
BS-2449R
BOSSBS-2449REA 752 EUR
BOSSBS-2449R
Anti-TNFRSF9 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TNFRSF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR