Anti-SLC16A3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Proveedor: Bioss
BS-2698R-A555
BOSSBS-2698R-A555EA 893 EUR
BOSSBS-2698R-A555
Anti-SLC16A3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticuerpos
Proton-linked monocarboxylate transporter. Catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC16A3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno SLC16A3
Nombre del antígeno Solute Carrier Family 16 Member A3
Environmentally Preferable
Sinónimos de antígenos MCT4|MCT-3|MCT 4|MCT-4|MCT 3|MCT3
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 555
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 9123
Isotipo IgG
InmunoQuímica Yes
Inmunógeno 406-460/465
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR