Anti-MCT4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Proveedor: Bioss
BS-2698R-A750
BOSSBS-2698R-A750EA 890 EUR
BOSSBS-2698R-A750
Anti-MCT4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
Proton-linked monocarboxylate transporter. catalyses the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate (By similarity).

Type: Primary
Antigen: MCT4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 401-465/465
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR