Anti-RGS5 Rabbit Polyclonal Antibody (Cy3®)

Proveedor: Bioss
BS-2794R-CY3
BOSSBS-2794R-CY3EA 900 EUR
BOSSBS-2794R-CY3
Anti-RGS5 Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
Inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. Binds to G(i)-alpha and G(o)-alpha, but not to G(s)-alpha.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RGS5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR