Anti-TNFAIP3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Proveedor: Bioss
BS-2803R-A680
BOSSBS-2803R-A680EA 890 EUR
BOSSBS-2803R-A680
Anti-TNFAIP3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticuerpos
Ubiquitin-editing enzyme that contains both ubiquitin ligase and deubiquitinase activities. Involved in immune and inflammatory responses signaled by cytokines, such as TNF-alpha and IL-1 beta, or pathogens via Toll-like receptors (TLRs) through terminating NF-kappa-B activity. Essential component of a ubiquitin-editing protein complex, comprising also RNF11, ITCH and TAX1BP1, that ensures the transient nature of inflammatory signaling pathways. In cooperation with TAX1BP1 promotes disassembly of E2-E3 ubiquitin protein ligase complexes in IL-1R and TNFR-1 pathways; affected are at least E3More Product Information