Anti-FOLH1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Proveedor: Bioss
BS-2899R-HRP
BOSSBS-2899R-HRPEA 1350 EUR
BOSSBS-2899R-HRP
Anti-FOLH1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Has both folate hydrolase and N-acetylated-alpha-linked-acidic dipeptidase (NAALADase) activity. Has a preference for tri-alpha-glutamate peptides. In the intestine, required for the uptake of folate. In the brain, modulates excitatory neurotransmission through the hydrolysis of the neuropeptide, N-aceylaspartylglutamate (NAAG), thereby releasing glutamate. Isoform PSM-4 and isoform PSM-5 would appear to be physiologically irrelevant. Involved in prostate tumor progression. Also exhibits a dipeptidyl-peptidase IV type activity. In vitro, cleaves Gly-Pro-AMC.
Recommended Dilutions: WesternMore Product Information