Anti-CHUK Rabbit Polyclonal Antibody (Cy3®)
Proveedor: Bioss
BS-3237R-CY3
BOSSBS-3237R-CY3EA 929 EUR
BOSSBS-3237R-CY3
Anti-CHUK Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
Serine kinase that plays an essential role in the NF-kappa-B signaling pathway which is activated by multiple stimuli such as inflammatory cytokines, bacterial or viral products, DNA damages or other cellular stresses. Acts as part of the canonical IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B on serine residues. These modifications allow polyubiquitination of the inhibitors and subsequent degradation by the proteasome. In turn, free NF-kappa-B is translocated into the nucleus and activates the transcription of hundreds of genesMore Product Information