Anti-GRIN2B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: Bioss
BS-3306R-FITC
BOSSBS-3306R-FITCEA 929 EUR
BOSSBS-3306R-FITC
Anti-GRIN2B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
NMDA receptor subtype of glutamate-gated ion channels with high calcium permeability and voltage-dependent sensitivity to magnesium. Mediated by glycine. In concert with DAPK1 at extrasynaptic sites, acts as a central mediator for stroke damage. Its phosphorylation at Ser-1303 by DAPK1 enhances synaptic NMDA receptor channel activity inducing injurious Ca2+ influx through them, resulting in an irreversible neuronal death (By similarity).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NMDAR2B
Clonality: Polyclonal
Clone:
Conjugation: FITC (More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR