Anti-THR927 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Proveedor: Bioss
BS-3445R-A350
BOSSBS-3445R-A350EA 921 EUR
BOSSBS-3445R-A350
Anti-THR927 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticuerpos
Mutations in this gene lead to tuberous sclerosis complex. Its gene product is believed to be a tumor suppressor and is able to stimulate specific GTPases. The protein associates with hamartin in a cytosolic complex, possibly acting as a chaperone for hamartin. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Tuberin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR