Anti-Tuberin Thr927 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Proveedor: Bioss
BS-3445R-A680
BOSSBS-3445R-A680EA 917 EUR
BOSSBS-3445R-A680
Anti-Tuberin Thr927 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticuerpos
Mutations in this gene lead to tuberous sclerosis complex. Its gene product is believed to be a tumor suppressor and is able to stimulate specific GTPases. The protein associates with hamartin in a cytosolic complex, possibly acting as a chaperone for hamartin. Alternative splicing results in multiple transcript variants encoding different isoforms.

Type: Primary
Antigen: Tuberin Thr927
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR