Anti-eNOS Ser1177 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Proveedor: Bioss
BS-3447R-A680
BOSSBS-3447R-A680EA 917 EUR
BOSSBS-3447R-A680
Anti-eNOS Ser1177 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticuerpos
Produces nitric oxide (NO) which is implicated in vascular smooth muscle relaxation through a cGMP-mediated signal transduction pathway. NO mediates vascular endothelial growth factor (VEGF)-induced angiogenesis in coronary vessels and promotes blood clotting through the activation of platelets. Isoform eNOS13C: Lacks eNOS activity, dominant-negative form that may down-regulate eNOS activity by forming heterodimers with isoform 1.

Type: Primary
Antigen: eNOS Ser1177
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR