Anti-RIPK2 Rabbit Polyclonal Antibody

Proveedor: Bioss
BS-3546R
BOSSBS-3546REA 725 EUR
BOSSBS-3546R
Anti-RIPK2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: RIPK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno RIPK2
Environmentally Preferable
Nombre del antígeno Receptor-interacting serine-threonine kinase 2
Sinónimos de antígenos CARD3|RICK|CARDIAK|receptor interacting serine/threonine kinase 2|GIG30|CCK|RIP2
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human,Rat,Mouse
Huésped Rabbit
Isotipo IgG
Western blot Yes
ELISA Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Adsorción cruzada none
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR