Anti-NCF/NCF4/p40phox Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Proveedor: Bioss
BS-3616R-A350
BOSSBS-3616R-A350EA 893 EUR
BOSSBS-3616R-A350
Anti-NCF/NCF4/p40phox Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticuerpos
NCF4 is a cytosolic oxidase associated protein. It is a component of the nicotinamide adenine dinucleotide phosphate oxidase, which mediates down regulation of NADPH oxidase through interactions with its SH3 domain. NCF4 is associated with p67 phox but is absent in patients with chronic granulomatous disease who lack p67 phox.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCF/NCF4/p40phox
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR