Anti-MLLT10 Rabbit Polyclonal Antibody (Cy3®)

Proveedor: Bioss
BS-3696R-CY3
BOSSBS-3696R-CY3EA 900 EUR
BOSSBS-3696R-CY3
Anti-MLLT10 Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
A chromosomal aberration involving MLLT10 is associated with acute leukemias. Translocation t(10;11)(p12;q23) with MLL/HRX. The result is a rogue activator protein.A chromosomal aberration involving MLLT10 is associated with diffuse histiocytic lymphomas. Translocation t(10;11)(p13;q14) with PICALM.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MLLT10
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR