Anti-GPR15 Rabbit Polyclonal Antibody (Cy3®)

Proveedor: Bioss
BS-3699R-CY3
BOSSBS-3699R-CY3EA 909 EUR
BOSSBS-3699R-CY3
Anti-GPR15 Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
GPR15 is a probable chemokine receptor,its expression has been reported in CD4(+) T lymphocytes, alveolar macrophages, and the basal surface of the small intestinal epithelium. ESTs have been isolated from normal brain and kidney cancer libraries.This gene encodes a G protein-coupled receptor that acts as a chemokine receptor for human immunodeficiency virus type 1 and 2. The encoded protein localizes to the cell membrane. [provided by RefSeq, Nov 2012].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPR15
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR