Anti-TP Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Proveedor: Bioss
BS-3809R-A647
BOSSBS-3809R-A647EA 893 EUR
BOSSBS-3809R-A647
Anti-TP Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticuerpos
In the presence of inorganic orthophosphate, the platelet-derived endothelial growth factor (PD-ECGF) thymidine phosphorylase (TP) (gliostatin) catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites.
Recommended Dilutions: IF(More Product Information