Anti-EPS8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Proveedor: Bioss
BS-3848R-A680
BOSSBS-3848R-A680EA 890 EUR
BOSSBS-3848R-A680
Anti-EPS8 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticuerpos
Signaling adapter that controls various cellular protrusions by regulating actin cytoskeleton dynamics and architecture. Depending on its association with other signal transducers, can regulate different processes. Together with SOS1 and ABI1, forms a trimeric complex that participates in transduction of signals from Ras to Rac by activating the Rac-specific guanine nucleotide exchange factor (GEF) activity. Acts as a direct regulator of actin dynamics by binding actin filaments and has both barbed-end actin filament capping and actin bundling activities depending on the context. DisplaysMore Product Information