Anti-NDUFS1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-3890R-A488
BOSSBS-3890R-A488EA 893 EUR
BOSSBS-3890R-A488
Anti-NDUFS1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone (By similarity). This is the largest subunit of complex I and it is a component of the iron-sulfur (IP) fragment of the enzyme. It may form part of the active site crevice where NADH is oxidized.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen:More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno NDUFS1
Nombre del antígeno NADH Dehydrogenase (Ubiquinone) Fe-S Protein 1, 75Kda (Nadh-Coenzyme Q Reductase)
Environmentally Preferable
Sinónimos de antígenos CI-75Kd|NADH-coenzyme Q reductase|PRO1304|mitochondrial respiratory chain|75kDa|NDUFS1|MGC26839|75-kD subunit|complex I|OTTHUMP00000206345|75kDa (NADH-coenzyme Q reductase)|NADH dehydrogenase (ubiquinone) Fe-S protein 1
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 488
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 4719
Isotipo IgG
InmunoQuímica Yes
Inmunógeno 90-130/727
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR