Anti-CYP24A1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-3899R-A488
BOSSBS-3899R-A488EA 893 EUR
BOSSBS-3899R-A488
Anti-CYP24A1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
Has a role in maintaining calcium homeostasis. Catalyzes the NADPH-dependent 24-hydroxylation of calcidiol (25-hydroxyvitamin D(3)) and calcitriol (1-alpha,25-dihydroxyvitamin D(3)). The enzyme can perform up to 6 rounds of hydroxylation of calcitriol leading to calcitroic acid. It also shows 23-hydroxylating activity leading to 1-alpha,25-dihydroxyvitamin D(3)-26,23-lactone as end product.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP24A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

ESPECIFICACIONES

Simbolo antígeno CYP24A1
Environmentally Preferable
Nombre del antígeno Cytochrome P450, Family 24, Subfamily A, Polypeptide 1
Sinónimos de antígenos CP24|HCAI|CYP24|P450-CC24
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 488
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 1591
Isotipo IgG
InmunoQuímica Yes
Inmunógeno 360-410/514
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR