Anti-SLC37A4 Rabbit Polyclonal Antibody (Cy3®)

Proveedor: Bioss
BS-4039R-CY3
BOSSBS-4039R-CY3EA 900 EUR
BOSSBS-4039R-CY3
Anti-SLC37A4 Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms a complex with glucose-6-phosphatase which is responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC37A4
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR