Anti-TBL1X Rabbit Polyclonal Antibody (Cy5®)

Proveedor: Bioss
BS-4219R-CY5
BOSSBS-4219R-CY5EA 900 EUR
BOSSBS-4219R-CY5
Anti-TBL1X Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
The protein encoded by this gene has sequence similarity with members of the WD40 repeat-containing protein family. The WD40 group is a large family of proteins, which appear to have a regulatory function. It is believed that the WD40 repeats mediate protein-protein interactions and members of the family are involved in signal transduction, RNA processing, gene regulation, vesicular trafficking, cytoskeletal assembly and may play a role in the control of cytotypic differentiation. This encoded protein is found as a subunit in corepressor SMRT (silencing mediator for retinoid and thyroidMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno TBL1X
Environmentally Preferable
Nombre del antígeno Transducin (beta)-like 1X-linked
Sinónimos de antígenos SMAP55|EBI|TBL1
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Cy5®
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 6907, 90665
Isotipo IgG
InmunoQuímica Yes
Inmunógeno 530-577/577
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR