Anti-TM7SF2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-4221R-A488
BOSSBS-4221R-A488EA 893 EUR
BOSSBS-4221R-A488
Anti-TM7SF2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
Involved in the conversion of lanosterol to cholesterol.Expressed in adult heart, brain, pancreas, lung, liver, skeletal muscle, kidney, ovary, prostate, and testis, but not detected in placenta, spleen, thymus, small intestine, colon (mucosal lining), or peripheral blood leukocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TM7SF2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR