Anti-VLDLR Rabbit Polyclonal Antibody (Cy5®)

Proveedor: Bioss
BS-4228R-CY5
BOSSBS-4228R-CY5EA 909 EUR
BOSSBS-4228R-CY5
Anti-VLDLR Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
Binds VLDL and transports it into cells by endocytosis. In order to be internalized, the receptor-ligand complexes must first cluster into clathrin-coated pits. Binding to Reelin induces tyrosine phosphorylation of Dab1 and modulation of Tau phosphorylation (By similarity).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: VLDLR
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR