Anti-SCNN1B Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Proveedor: Bioss
BS-4252R-A555
BOSSBS-4252R-A555EA 893 EUR
BOSSBS-4252R-A555
Anti-SCNN1B Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticuerpos
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Plays an essential role in electrolyte and blood pressure homeostasis, but also in airway surface liquid homeostasis, which is important for proper clearance of mucus. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCNN1B
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR