Anti-SCNN1B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: Bioss
BS-4252R-FITC
BOSSBS-4252R-FITCEA 900 EUR
BOSSBS-4252R-FITC
Anti-SCNN1B Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Plays an essential role in electrolyte and blood pressure homeostasis, but also in airway surface liquid homeostasis, which is important for proper clearance of mucus. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCNN1B
Clonality:More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno SCNN1B
Nombre del antígeno Sodium Channel Nonvoltage-gated 1 Beta
Sinónimos de antígenos ENaCb|BESC1|ENaCbeta|SCNEB
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado FITC (Fluorescein Isothiocyanate)
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 6338
Isotipo IgG
InmunoQuímica Yes
Inmunógeno 430-480/685
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR