Anti-CUEDC2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Proveedor: Bioss
BS-4608R-A750
BOSSBS-4608R-A750EA 890 EUR
BOSSBS-4608R-A750
Anti-CUEDC2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
Down-regulates ESR1 protein levels through the ubiquitination-proteasome pathway, regardless of the presence of 17 beta-estradiol. Also involved in 17 beta-estradiol-induced ESR1 degradation. Controls PGR protein levels through a similar mechanism.

Type: Primary
Antigen: CUEDC2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-287/287
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR