Anti-MAL Rabbit Polyclonal Antibody (Cy5®)

Proveedor: Bioss
BS-4835R-CY5
BOSSBS-4835R-CY5EA 900 EUR
BOSSBS-4835R-CY5
Anti-MAL Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
Could be an important component in vesicular trafficking cycling between the Golgi complex and the apical plasma membrane. Could be involved in myelin biogenesis and/or myelin function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAL
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR