Anti-ACADL Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Proveedor: Bioss
BS-5015R-A647
BOSSBS-5015R-A647EA 893 EUR
BOSSBS-5015R-A647
Anti-ACADL Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticuerpos
The protein encoded by this gene belongs to the acyl-CoA dehydrogenase family, which is a family of mitochondrial flavoenzymes involved in fatty acid and branched chain amino-acid metabolism. This protein is one of the four enzymes that catalyze the initial step of mitochondrial beta-oxidation of straight-chain fatty acid. Defects in this gene are the cause of long-chain acyl-CoA dehydrogenase (LCAD) deficiency, leading to nonketotic hypoglycemia. [provided by RefSeq].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACADL
Clonality: Polyclonal
Clone:
Conjugation: AlexaMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR