Anti-ACADL Rabbit Polyclonal Antibody (Cy5.5®)

Proveedor: Bioss
BS-5015R-CY5.5
BOSSBS-5015R-CY5.5EA 900 EUR
BOSSBS-5015R-CY5.5
Anti-ACADL Rabbit Polyclonal Antibody (Cy5.5®)
Anticuerpos
The protein encoded by this gene belongs to the acyl-CoA dehydrogenase family, which is a family of mitochondrial flavoenzymes involved in fatty acid and branched chain amino-acid metabolism. This protein is one of the four enzymes that catalyze the initial step of mitochondrial beta-oxidation of straight-chain fatty acid. Defects in this gene are the cause of long-chain acyl-CoA dehydrogenase (LCAD) deficiency, leading to nonketotic hypoglycemia. [provided by RefSeq].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACADL
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno ACADL
Nombre del antígeno Acyl-CoA Dehydrogenase Long Chain
Environmentally Preferable
Sinónimos de antígenos ACAD4|LCAD
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Cy5.5®
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 33
Isotipo IgG
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR