Anti-AHCY Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Proveedor: Bioss
BS-5029R-A350
BOSSBS-5029R-A350EA 901 EUR
BOSSBS-5029R-A350
Anti-AHCY Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticuerpos
Adenosylhomocysteine is a competitive inhibitor of S-adenosyl-L-methionine-dependent methyl transferase reactions; therefore adenosylhomocysteinase may play a key role in the control of methylations via regulation of the intracellular concentration of adenosylhomocysteine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AHCY
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno AHCY
Nombre del antígeno Adenosylhomocysteinase
Environmentally Preferable
Sinónimos de antígenos adoHcyase|SAHH
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 350
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 191
Isotipo IgG
InmunoQuímica Yes
Inmunógeno 20-70/432
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR