Anti-TSC2 Rabbit Polyclonal Antibody (Cy7®)
Proveedor: Bioss
BS-5609R-CY7
BOSSBS-5609R-CY7EA 1350 EUR
BOSSBS-5609R-CY7
Anti-TSC2 Rabbit Polyclonal Antibody (Cy7®)
Anticuerpos
Tuberin, or TSC2 (Tuberous sclerosis complex), is implicated as a tumor suppressor. It may function in vesicular transport, and may also play a role in the regulation of cell growth arrest and in the regulation of transcription mediated by steroid receptors. Interaction between hamartin (TSC1) and tuberin may facilitate vesicular docking. It specifically stimulates the intrinsic GTPase activity of the Ras related protein RAP1A and RAB5, suggesting a possible mechanism for its role in regulating cellular growth. Mutations in tuberin lead to constitutive activation of RAP1A in tumors. At leastMore Product Information