Anti-RAB24 Rabbit Polyclonal Antibody (Cy5.5®)

Proveedor: Bioss
BS-5690R-CY5.5
BOSSBS-5690R-CY5.5EA 1350 EUR
BOSSBS-5690R-CY5.5
Anti-RAB24 Rabbit Polyclonal Antibody (Cy5.5®)
Anticuerpos
A major contributor to cellular homeostasis is the ability of the cell to strike a balance between the formation and degradation/removal of its cellular components. This process of internal cellular turn-over is called autophagy (self-eating), and is facilitated by a pathway of around 16 interacting proteins in the human. The GTPase Rab24 is thought to be involved in the regulation of vesicular transport associated with autophagy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAB24
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR