Anti-SCNN1B Thr615 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Proveedor: Bioss
BS-5701R-A680
BOSSBS-5701R-A680EA 921 EUR
BOSSBS-5701R-A680
Anti-SCNN1B Thr615 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticuerpos
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Plays an essential role in electrolyte and blood pressure homeostasis, but also in airway surface liquid homeostasis, which is important for proper clearance of mucus. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.

Type: Primary
Antigen: SCNN1B Thr615
Clonality: Polyclonal
Clone:
Conjugation: ALEXAMore Product Information
Order Now

ESPECIFICACIONES

Nombre del antígeno SCNN1B Thr615
Environmentally Preferable
Sinónimos de antígenos Nonvoltage-gated sodium channel 1 subunit beta|Beta-NaCH|SCNN1B|ENaCb|Epithelial Na(+) channel subunit beta|BESC1|ENaCbeta|Beta-ENaC|SCNEB|Amiloride-sensitive sodium channel subunit beta
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 680
Reactividad Human
Huésped Rabbit
ID del gen 6338
Isotipo IgG
Western blot Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Modificación Phosphorylation (Thr615)
Inmunógeno KLH conjugated synthetic phosphopeptide derived from human SCNN1B around the phosphorylation site of Thr615
Forma Liquid
Molecular weight 73 kDa
Purification Purified by Protein A
Concentración 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Temperatura de almacenamiento Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR