Anti-FAM13B1 Rabbit Polyclonal Antibody (Cy3®)

Proveedor: Bioss
BS-5757R-CY3
BOSSBS-5757R-CY3EA 900 EUR
BOSSBS-5757R-CY3
Anti-FAM13B1 Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
FAM13B1

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM13B1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR