Anti-ADAMTSL3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Proveedor: Bioss
BS-5863R-A750
BOSSBS-5863R-A750EA 890 EUR
BOSSBS-5863R-A750
Anti-ADAMTSL3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
Expressed in epithelial cells of the colon, fallopian tube, skin, breast, prostate, epididymis, liver, pancreatic islets and bile ducts, as well as by vascular endothelial cells, smooth muscle cells, fibroblasts, cortical and ganglionic neurons and cardiac myocytes. Also expressed by malignant epithelial cells in colon cancer, as well as breast, prostate, renal and skin tumors. Expression is significantly reduced in colon cancer compared to normal colon.

Type: Primary
Antigen: ADAMTSL3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 751-850/1691
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR