Anti-ADAMTSL3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Bioss
BS-5863R-HRP
BOSSBS-5863R-HRPEA 900 EUR
BOSSBS-5863R-HRP
Anti-ADAMTSL3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Expressed in epithelial cells of the colon, fallopian tube, skin, breast, prostate, epididymis, liver, pancreatic islets and bile ducts, as well as by vascular endothelial cells, smooth muscle cells, fibroblasts, cortical and ganglionic neurons and cardiac myocytes. Also expressed by malignant epithelial cells in colon cancer, as well as breast, prostate, renal and skin tumors. Expression is significantly reduced in colon cancer compared to normal colon.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ADAMTSL3
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR