Anti-PRDM2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Proveedor: Bioss
BS-6062R-A555
BOSSBS-6062R-A555EA 893 EUR
BOSSBS-6062R-A555
Anti-PRDM2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticuerpos
S-adenosyl-L-methionine-dependent histone methyltransferase that specifically methylates 'Lys-9' of histone H3. May function as a DNA-binding transcription factor. Binds to the macrophage-specific TPA-responsive element (MTE) of the HMOX1 (heme oxygenase 1) gene and may act as a transcriptional activator of this gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRDM2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno PRDM2
Nombre del antígeno PR Domain Zinc Finger Protein 2
Environmentally Preferable
Sinónimos de antígenos RIZ2|KMT8|MTB-ZF|RIZ1|RIZ|HUMHOXY1
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 555
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 7799
Isotipo IgG
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR