Anti-HRAS Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Proveedor: Bioss
BS-6165R-A350
BOSSBS-6165R-A350EA 893 EUR
BOSSBS-6165R-A350
Anti-HRAS Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticuerpos
The KRAS gene encodes the human cellular homolog of a transforming gene isolated from the Kirsten rat sarcoma virus. The RAS proteins are GDP/GTP-binding proteins that act as intracellular signal transducers. The most well-studied members of the RAS (derived from 'RAt Sarcoma' virus) gene family include KRAS, HRAS, and NRAS. These genes encode immunologically related proteins with a molecular mass of 21 kD and are homologs of rodent sarcoma virus genes that have transforming abilities. While these wildtype cellular proteins in humans play a vital role in normal tissue signaling, includingMore Product Information