Anti-TMPRSS2 Rabbit Polyclonal Antibody (Cy5®)
Proveedor: Bioss
BS-6285R-CY5
BOSSBS-6285R-CY5EA 900 EUR
BOSSBS-6285R-CY5
Anti-TMPRSS2 Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
Serine protease that proteolytically cleaves and activates the viral spike glycoproteins which facilitate virus-cell membrane fusions; spike proteins are synthesized and maintained in precursor intermediate folding states and proteolysis permits the refolding and energy release required to create stable virus-cell linkages and membrane coalescence. Facilitates human SARS coronavirus (SARS-CoV) infection via two independent mechanisms, proteolytic cleavage of ACE2, which might promote viral uptake, and cleavage of coronavirus spike glycoprotein which activates the glycoprotein for cathepsin L-More Product Information