Anti-Quiescin Q6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Proveedor: Bioss
BS-6394R-A555
BOSSBS-6394R-A555EA 893 EUR
BOSSBS-6394R-A555
Anti-Quiescin Q6 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticuerpos
This gene encodes a protein that contains domains of thioredoxin and ERV1, members of two long-standing gene families. The gene expression is induced as fibroblasts begin to exit the proliferative cycle and enter quiescence, suggesting that this gene plays an important role in growth regulation. Two transcript variants encoding two different isoforms have been found for this gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Quiescin Q6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR