Anti-CA8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: Bioss
BS-6516R-FITC
BOSSBS-6516R-FITCEA 900 EUR
BOSSBS-6516R-FITC
Anti-CA8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
The protein encoded by this gene was initially named CA-related protein because of sequence similarity to other known carbonic anhydrase genes. However, the gene product lacks carbonic anhydrase activity (i.e., the reversible hydration of carbon dioxide). The gene product continues to carry a carbonic anhydrase designation based on clear sequence identity to other members of the carbonic anhydrase gene family. The absence of CA8 gene transcription in the cerebellum of the lurcher mutant in mice with a neurologic defect suggests an important role for this acatalytic form.

RecommendedMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno CA8
Nombre del antígeno Carbonic Anhydrase 8
Environmentally Preferable
Sinónimos de antígenos CALS|CA-VIII|CARP|CAMRQ3
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado FITC (Fluorescein Isothiocyanate)
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 767
Isotipo IgG
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR